Learn More
Research Products International Corp Beef Extract, 5 Kilograms
SDP
Catalog No. 501368103
Encompass_Preferred
Beef Extract is used as a nutrient source in various culture media.
Catalog No. 50-136-8103 Supplier Research Products International Corp Supplier No. B101505000.0
Beef Extract is used as a nutrient source in various culture media.