Learn More
Alkali Scientific Beef Extract, 500 Grams
SDP
Catalog No. 5000661667
Encompass_Preferred
Beef Extract, 500 Grams is a nutrient-rich extract commonly used as a supplement in microbiological growth media. Beef extract provides essential growth factors, vitamins, amino acids, and nitrogenous compounds that support the growth of a wide range of microorganisms. This 500g pack size is ideal for laboratories that prepare bacterial or fungal culture media in large quantities. The extract is widely used in the preparation of nutrient-rich media for growing bacteria, fungi, and yeast strains in clinical, research, and industrial applications. It is a critical component in many microbiological protocols for isolation, identification, and propagation of microorganisms.
Catalog No. 50-006-61667 Supplier Alkali Scientific Supplier No. BEE001CP
Beef Extract, 500 Grams is a nutrient-rich extract commonly used as a supplement in microbiological growth media. Beef extract provides essential growth factors, vitamins, amino acids, and nitrogenous compounds that support the growth of a wide range of microorganisms. This 500g pack size is ideal for laboratories that prepare bacterial or fungal culture media in large quantities. The extract is widely used in the preparation of nutrient-rich media for growing bacteria, fungi, and yeast strains in clinical, research, and industrial applications. It is a critical component in many microbiological protocols for isolation, identification, and propagation of microorganisms.