Learn More
Bioworld Beef Extract, 500 g
Catalog No. 50254275
Encompass_Preferred
Beef Extract (Powder)Recommended for use as a nutrient at 0.3-0.5% concentration in various culture mediaCan be used in fermentation studies as it is free of fermentable substratepH: 7.0Dehydrated Appearance: cream to tan, homogeneous powderPrepared Appearance: yellow to tanSoluble in water
Catalog No. 50-254-275 Supplier Bioworld Supplier No. 306200081
Beef Extract (Powder)Recommended for use as a nutrient at 0.3-0.5% concentration in various culture mediaCan be used in fermentation studies as it is free of fermentable substratepH: 7.0Dehydrated Appearance: cream to tan, homogeneous powderPrepared Appearance: yellow to tanSoluble in water