Learn More
NEOGEN Beef Extract Powder, 5 kg
SDP
Catalog No. 502015742
Encompass_Preferred
Beef Extract Powder is a dehydrated extract of bovine tissue for use in preparing microbiological culture media in a laboratory setting. Beef Extract Powder is not intended for use in the diagnosis of disease or other conditions in humans.Beef Extract Powder is prepared and standardized for use in microbiological culture media, where it is generally used to replace infusion of meat. Culture media containing Beef Extract Powder are recommended for use in bacteriological examination of water, milk, and other materials, where uniform composition of media is important. Beef Extract Powder is relied upon for biochemical studies, particularly fermentation reactions because of its independence from fermentable substances. Several media containing Beef Extract Powder are recommended in standard methods for multiple application.For more information visit https://www.neogen.com/solutions/Microbiology/beef-extract-powder
Catalog No. 50-201-5742 Supplier NEOGEN Supplier No. 700003511
Beef Extract Powder is a dehydrated extract of bovine tissue for use in preparing microbiological culture media in a laboratory setting. Beef Extract Powder is not intended for use in the diagnosis of disease or other conditions in humans.Beef Extract Powder is prepared and standardized for use in microbiological culture media, where it is generally used to replace infusion of meat. Culture media containing Beef Extract Powder are recommended for use in bacteriological examination of water, milk, and other materials, where uniform composition of media is important. Beef Extract Powder is relied upon for biochemical studies, particularly fermentation reactions because of its independence from fermentable substances. Several media containing Beef Extract Powder are recommended in standard methods for multiple application.For more information visit https://www.neogen.com/solutions/Microbiology/beef-extract-powder