Learn More
Electron Microscopy Sciences BEEM Block Storage
SDP
Supplier:  Electron Microscopy Sciences 6992201
Encompass_Preferred
A two tone color coded plastic box with insert, which allows for the storage of up to 10 blocks in the numbered cavities.
Catalog No. 50-246-43