Learn More
Electron Microscopy Sciences BEEM Block Storage
SDP
Catalog No. 5024644
Encompass_Preferred
A two tone color coded plastic box with insert, which allows for the storage of up to 10 blocks in the numbered cavities.
Catalog No. 50-246-44 Supplier Electron Microscopy Sciences Supplier No. 6992205
A two tone color coded plastic box with insert, which allows for the storage of up to 10 blocks in the numbered cavities.