Learn More
Electron Microscopy Sciences BEEM Blocklock Flat Embedment, 20 Positions
SDP
Catalog No. 5030477
Encompass
Specimen blocks are safely stored in Blocklock storage modules, a component in the BEEM Specimen Storage System 2000. All Blocklocks can be inserted in a Plastic Protective Box or, by utilizing the adhesive feature, mounted in combination with other Blocklock modules on the BEEM mounting panel that fits in a standard " D-Type" 3-ring binder. Block snaps into cavities so it cannot be inadvertently dislodged. Number and letter identify each cavity. Measures 2-7⁄8"L x 2"W x 1/4"H (74 x 50 x 8mm)
Catalog No. 50-304-77 Supplier Electron Microscopy Sciences Supplier No. 69959
Specimen blocks are safely stored in Blocklock storage modules, a component in the BEEM Specimen Storage System 2000. All Blocklocks can be inserted in a Plastic Protective Box or, by utilizing the adhesive feature, mounted in combination with other Blocklock modules on the BEEM mounting panel that fits in a standard " D-Type" 3-ring binder. Block snaps into cavities so it cannot be inadvertently dislodged. Number and letter identify each cavity. Measures 2-7⁄8"L x 2"W x 1/4"H (74 x 50 x 8mm)