Learn More
Electron Microscopy Sciences BEEM Blocklock Storage for #69957
SDP
Catalog No. 5030481
Encompass
Specimen blocks are safely stored in Blocklock storage modules, a component in the BEEM Specimen Storage System 2000. All Blocklocks can be inserted in a Plastic Protective Box or, by utilizing the adhesive feature, mounted in combination with other Blocklock modules on the BEEM mounting panel that fits in a standard " D-Type" 3-ring binder. Heavy-duty, two tone plastic box supplied with indexed translucent labels. Two-tone blue/clear, overall measurement: 4 3/4"(L) x 3 3/4"(W) x1/2"(H) (120 x 94 x 13mm)
Catalog No. 50-304-81 Supplier Electron Microscopy Sciences Supplier No. 69957B
Specimen blocks are safely stored in Blocklock storage modules, a component in the BEEM Specimen Storage System 2000. All Blocklocks can be inserted in a Plastic Protective Box or, by utilizing the adhesive feature, mounted in combination with other Blocklock modules on the BEEM mounting panel that fits in a standard " D-Type" 3-ring binder. Heavy-duty, two tone plastic box supplied with indexed translucent labels. Two-tone blue/clear, overall measurement: 4 3/4"(L) x 3 3/4"(W) x1/2"(H) (120 x 94 x 13mm)