Learn More
Electron Microscopy Sciences BEEM Blocklock Storage Box for #69956
SDP
Catalog No. 5030483
Encompass
Specimen blocks are safely stored in Blocklock storage modules, a component in the BEEM Specimen Storage System 2000. All Blocklocks can be inserted in a Plastic Protective Box or, by utilizing the adhesive feature, mounted in combination with other Blocklock modules on the BEEM mounting panel that fits in a standard " D-Type" 3-ring binder. Heavy-duty, two tone plastic box supplied with indexed translucent labels for top or bottom of the box. Storage for #69956
Catalog No. 50-304-83 Supplier Electron Microscopy Sciences Supplier No. 69956B
Specimen blocks are safely stored in Blocklock storage modules, a component in the BEEM Specimen Storage System 2000. All Blocklocks can be inserted in a Plastic Protective Box or, by utilizing the adhesive feature, mounted in combination with other Blocklock modules on the BEEM mounting panel that fits in a standard " D-Type" 3-ring binder. Heavy-duty, two tone plastic box supplied with indexed translucent labels for top or bottom of the box. Storage for #69956