Learn More
Electron Microscopy Sciences BEEM Capsule Holder, Clear
SDP
Catalog No. 5029277
Encompass_Preferred
If microwaving or UV polymerizing of your embedding media these opaque BEEM capsule holders are perfect. Made from Polypropylene. All embedding capsules size 00 fit it.
Catalog No. 50-292-77 Supplier Electron Microscopy Sciences Supplier No. 6991506
If microwaving or UV polymerizing of your embedding media these opaque BEEM capsule holders are perfect. Made from Polypropylene. All embedding capsules size 00 fit it.