Learn More
Electron Microscopy Sciences BEEM Capsule Holder, Clear
SDP
Supplier:  Electron Microscopy Sciences 6991501
Encompass_Preferred
If microwaving or UV polymerizing of your embedding media these opaque BEEM capsule holders are perfect. Made from Polypropylene. All embedding capsules size 00 fit it.
Catalog No. 50-292-76