Learn More
Electron Microscopy Sciences BEEM Capsule Holder, Size 00
SDP
Supplier:  Electron Microscopy Sciences 6991606
Encompass_Preferred
These holders are used to hold capsules upright during the resin embedding stage. The cavities (22) allow for illumination from the bottom of the holder for specimen orientation. Holds size "00", conical, and bottle neck capsules.
Size: Size 00
Catalog No. 50-246-38