Learn More
Electron Microscopy Sciences BEEM Capsule Holder, Size 00
SDP
Catalog No. 5024635
Encompass_Preferred
These holders are used to hold capsules upright during the resin embedding stage. The cavities (22) allow for illumination from the bottom of the holder for specimen orientation. Holds size "00", conical, and bottle neck capsules.
Size: Size 00
Catalog No. 50-246-35 Supplier Electron Microscopy Sciences Supplier No. 6991601
These holders are used to hold capsules upright during the resin embedding stage. The cavities (22) allow for illumination from the bottom of the holder for specimen orientation. Holds size "00", conical, and bottle neck capsules.
Size: Size 00