Learn More
Electron Microscopy Sciences BEEM Capsule Holder White, Size 3
SDP
Catalog No. 501927445
Encompass_Preferred
These holders are used to hold capsules upright during the resin embedding stage. The cavities (22) allow for illumination from the bottom of the holder for specimen orientation. Holds size "3" capsules.
Size: Size 3
Catalog No. 50-192-7445 Supplier Electron Microscopy Sciences Supplier No. 69917013
These holders are used to hold capsules upright during the resin embedding stage. The cavities (22) allow for illumination from the bottom of the holder for specimen orientation. Holds size "3" capsules.
Size: Size 3