Learn More
Electron Microscopy Sciences BEEM Capsule Holder White, Size 3
SDP
Supplier:  Electron Microscopy Sciences 69917063
Encompass_Preferred
These holders are used to hold capsules upright during the resin embedding stage. The cavities (22) allow for illumination from the bottom of the holder for specimen orientation. Holds size "3" capsules.
Size: Size 3
Catalog No. 50-192-7444