Learn More
Electron Microscopy Sciences BEEM Dial-A-Grid & Block Storage
SDP
Catalog No. 50930856
Encompass
Two tone color coded plastic box with insert which has 24 letter-labeled crossed slots and 2 cavities for block storage. A rotating protection plate covers the slots and allows for exposure of one grid at a time. There are two additional cavities for block storage. BEEM Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
Catalog No. 50-930-856 Supplier Electron Microscopy Sciences Supplier No. 7114910
Two tone color coded plastic box with insert which has 24 letter-labeled crossed slots and 2 cavities for block storage. A rotating protection plate covers the slots and allows for exposure of one grid at a time. There are two additional cavities for block storage. BEEM Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.