Learn More
Electron Microscopy Sciences BEEM Dial-A-Grid Storage Box
SDP
Supplier:  Electron Microscopy Sciences 7114805
Encompass_Preferred
A two tone color coded plastic box with insert which has 24 letter-labeled crossed slots, where the grids can be stored. A rotating protection plate covers the slots and allows for exposure of one grid at a time. BEEM Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
Catalog No. 50-930-852