Learn More
Electron Microscopy Sciences BEEM Embedding Capsule Bottle Neck Tip
SDP
Catalog No. 5024653
Encompass_Preferred
Specially shaped capsules are designed to overcome commonly encountered embedding problems; such as, small or thin elongated specimens and powder samples. These capsules are also good for centrifuging precipitated matters and allows for less trimming. BEEM Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc. Bottle neck capsule produces a 7.9mm OD block with a hemihyperpoloid (rather than faced) tip.
Catalog No. 50-246-53 Supplier Electron Microscopy Sciences Supplier No. 6991205
Specially shaped capsules are designed to overcome commonly encountered embedding problems; such as, small or thin elongated specimens and powder samples. These capsules are also good for centrifuging precipitated matters and allows for less trimming. BEEM Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc. Bottle neck capsule produces a 7.9mm OD block with a hemihyperpoloid (rather than faced) tip.