Learn More
Electron Microscopy Sciences BEEM Embedding Capsule Conical Tip
SDP
Catalog No. 5024655
Encompass_Preferred
Specially shaped capsules are designed to overcome commonly encountered embedding problems; such as, small or thin elongated specimens and powder samples. These capsules are also good for centrifuging precipitated matters and allows for less trimming. BEEM Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc. Conical capsule produces a 7.9mm OD block with a conical (rather than faced) tip.
Catalog No. 50-246-55 Supplier Electron Microscopy Sciences Supplier No. 6991310
Specially shaped capsules are designed to overcome commonly encountered embedding problems; such as, small or thin elongated specimens and powder samples. These capsules are also good for centrifuging precipitated matters and allows for less trimming. BEEM Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc. Conical capsule produces a 7.9mm OD block with a conical (rather than faced) tip.