Learn More
Electron Microscopy Sciences BEEM Embedding Capsules size 00, 8 mm I.D.
SDP
Catalog No. 5024869
Encompass_Preferred
Polyethylene embedding capsules with 1x1 mm face at the tip of a truncated pyramid, each with a hinged cap.
I.D.: 8 mm
Catalog No. 50-248-69 Supplier Electron Microscopy Sciences Supplier No. 70010B
Polyethylene embedding capsules with 1x1 mm face at the tip of a truncated pyramid, each with a hinged cap.
I.D.: 8 mm