Learn More
Electron Microscopy Sciences BEEM Embedding Capsules, Size 3
SDP
Catalog No. 5024649
Encompass_Preferred
These capsules, made from polyethylene, produce blocks with a 5.6 mm O.D. and a pyramid tip. Comes complete with a hinged cap.
Catalog No. 50-246-49 Supplier Electron Microscopy Sciences Supplier No. 6991001
These capsules, made from polyethylene, produce blocks with a 5.6 mm O.D. and a pyramid tip. Comes complete with a hinged cap.