Learn More
Electron Microscopy Sciences BEEM Flat Embedding Mold
SDP
Catalog No. 50930537
Encompass_Preferred
Molded from polyethylene. The critical surfaces of the mold are transparent, facilitating accurate specimen orientation through bottom illumination. Molds are reusable and each mold is supplied with a protective box.
Catalog No. 50-930-537 Supplier Electron Microscopy Sciences Supplier No. 7090412
Molded from polyethylene. The critical surfaces of the mold are transparent, facilitating accurate specimen orientation through bottom illumination. Molds are reusable and each mold is supplied with a protective box.