Learn More
Electron Microscopy Sciences BEEM Mounting Panel with Clear Cover
SDP
Catalog No. 5030487
Encompass
Panel for BEEM Dial- A-Grid, Block Storage Modules or grid storage boxes not exceeding 3⁄8". Attach with double sided adhesive tape. Measures: 10 5⁄8"(L) x 7.5"(W) x 3⁄8"(H)
Catalog No. 50-304-87 Supplier Electron Microscopy Sciences Supplier No. 69950
Panel for BEEM Dial- A-Grid, Block Storage Modules or grid storage boxes not exceeding 3⁄8". Attach with double sided adhesive tape. Measures: 10 5⁄8"(L) x 7.5"(W) x 3⁄8"(H)