Learn More
Electron Microscopy Sciences BEEM Opaque Capsule Holder Size 3 Capsule
SDP
Catalog No. 501901629
Encompass_Preferred
If microwaving or UV polymerizing of your embedding media these opaque BEEM capsule holders are perfect. Made from Polypropylene. All embedding capsules size 00 fit it.
Catalog No. 50-190-1629 Supplier Electron Microscopy Sciences Supplier No. 69915033
If microwaving or UV polymerizing of your embedding media these opaque BEEM capsule holders are perfect. Made from Polypropylene. All embedding capsules size 00 fit it.