Learn More
MSC Diversey Glass Cleaner, 4 lb, Unscented, Powder, 2/CT
Catalog No. 502819752
Encompass
Diversey Beer Clean Glass Cleaner is a powder-form cleaning agent designed for industrial and institutional use, specifically for hand washing beverage glasses. It is intended to be diluted prior to use and contains a blend of cleaning agents.
  • Container type: Bucket
  • Container size: 4 lb
  • Unscented
  • Application: Beverage glass
  • Chemical composition includes sodium citrate, sodium sesquicarbonate, sodium sulfate, tetrasodium salt of EDTA, and linear alkyl benzene sulfonates
  • Completely soluble
  • Dilution pH of 9.45 at 1:768
Catalog No. 50-281-9752 Supplier MSC Supplier No. 20467783
Diversey Beer Clean Glass Cleaner is a powder-form cleaning agent designed for industrial and institutional use, specifically for hand washing beverage glasses. It is intended to be diluted prior to use and contains a blend of cleaning agents.
  • Container type: Bucket
  • Container size: 4 lb
  • Unscented
  • Application: Beverage glass
  • Chemical composition includes sodium citrate, sodium sesquicarbonate, sodium sulfate, tetrasodium salt of EDTA, and linear alkyl benzene sulfonates
  • Completely soluble
  • Dilution pH of 9.45 at 1:768