Learn More
Medchemexpress LLC Beeswax | 8012-89-3 | MFCD00132754 | Variable; no single molecular weight (approx. 900 g/mol) | No single chemical formula - complex mixture of esters, fatty acids, and hydrocarbons | 5 KG
SDP
Catalog No. 5000282999
Encompass_Preferred
Beeswax is a natural wax mixture of esters, fatty acids, and hydrocarbons supplied as a CP-grade biochemical reagent for laboratory and life-science use. It is solid at room temperature and easy to melt, making it suitable for formulation, coating, embedding, and sample preparation. Documentation such as an SDS and a certificate of analysis is provided for quality control.
  • Natural mixture of esters, fatty acids, and hydrocarbons.
  • Chemical pure (CP) grade suitable for laboratory use.
  • Solid at room temperature; easy to melt and cast.
  • Useful for formulation, coating, embedding, and sample preparation.
  • Provided with SDS and certificate of analysis for quality control.
  • Available in multiple pack sizes, including bulk 5 KG packaging.
Catalog No. 50-002-82999 Supplier Medchemexpress LLC Supplier No. HYW1339745KG
Beeswax is a natural wax mixture of esters, fatty acids, and hydrocarbons supplied as a CP-grade biochemical reagent for laboratory and life-science use. It is solid at room temperature and easy to melt, making it suitable for formulation, coating, embedding, and sample preparation. Documentation such as an SDS and a certificate of analysis is provided for quality control.
  • Natural mixture of esters, fatty acids, and hydrocarbons.
  • Chemical pure (CP) grade suitable for laboratory use.
  • Solid at room temperature; easy to melt and cast.
  • Useful for formulation, coating, embedding, and sample preparation.
  • Provided with SDS and certificate of analysis for quality control.
  • Available in multiple pack sizes, including bulk 5 KG packaging.