Learn More
Medchemexpress LLC Befiradol hydrochloride | 2436760-81-3 | 99.93% | 430.32 | 25 MG
SDP
Catalog No. 5000362783
Encompass_Preferred
Befiradol hydrochloride is a selective 5-HT1A receptor agonist, suitable for research use only.
  • Reduces activity of dorsal raphe serotonergic neurons
  • Increases discharge rate of mPFC pyramidal neurons
  • Decreases extracellular 5-HT in the hippocampus and mPFC
  • Increases extracellular DA in mPFC
Catalog No. 50-003-62783 Supplier Medchemexpress LLC Supplier No. 25MGHY14785A
Befiradol hydrochloride is a selective 5-HT1A receptor agonist, suitable for research use only.
  • Reduces activity of dorsal raphe serotonergic neurons
  • Increases discharge rate of mPFC pyramidal neurons
  • Decreases extracellular 5-HT in the hippocampus and mPFC
  • Increases extracellular DA in mPFC