Learn More
Medchemexpress LLC Befiradol hydrochloride | 2436760-81-3 | MFCD31731608 | 99.9% | 430.32 g/mol | C20H23Cl2F2N3O | 10 MG
SDP
Catalog No. 5000286970
Encompass_Preferred
Befiradol hydrochloride is a selective 5-HT1A receptor agonist provided for research use. It modulates serotonergic and dopaminergic activity in cortical regions and is used in preclinical pharmacology studies to investigate 5-HT1A-mediated mechanisms and models of neurological disorders. The material is supplied with supporting documentation for analytical and safety purposes.
  • Selective 5-HT1A receptor agonist for receptor pharmacology studies.
  • Demonstrated activity on dorsal raphe and medial prefrontal cortex neurotransmission.
  • High chemical purity suitable for analytical and in vivo research.
  • Supplied with certificate of analysis and safety data sheet.
  • Available in multiple pack sizes for small-scale and bulk use.
  • Not for human or clinical use; for research use only.
Catalog No. 50-002-86970 Supplier Medchemexpress LLC Supplier No. HY14785A10MG
Befiradol hydrochloride is a selective 5-HT1A receptor agonist provided for research use. It modulates serotonergic and dopaminergic activity in cortical regions and is used in preclinical pharmacology studies to investigate 5-HT1A-mediated mechanisms and models of neurological disorders. The material is supplied with supporting documentation for analytical and safety purposes.
  • Selective 5-HT1A receptor agonist for receptor pharmacology studies.
  • Demonstrated activity on dorsal raphe and medial prefrontal cortex neurotransmission.
  • High chemical purity suitable for analytical and in vivo research.
  • Supplied with certificate of analysis and safety data sheet.
  • Available in multiple pack sizes for small-scale and bulk use.
  • Not for human or clinical use; for research use only.