Learn More
ATHENA ES BEGM Medium
Catalog No. 5000158159
Encompass_Preferred
BEGM (Bronchial Epithelial Growth Medium) is supplied as a complete serum-free medium comprised of essential amino acids, vitamins, inorganic salts, trace elements and growth factors optimized for culturing initial harvest of human airway epithelial cells and for expansion of established cell lines. BEGM is derived from LHC Basal Medium which is supplemented with growth factors and supplements that eliminate the need for serum.
Catalog No. 50-001-58159 Supplier ATHENA ES Supplier No. 0451
BEGM (Bronchial Epithelial Growth Medium) is supplied as a complete serum-free medium comprised of essential amino acids, vitamins, inorganic salts, trace elements and growth factors optimized for culturing initial harvest of human airway epithelial cells and for expansion of established cell lines. BEGM is derived from LHC Basal Medium which is supplemented with growth factors and supplements that eliminate the need for serum.