Learn More
Waters Corp Waters UPLC Column, Glycan BEH Amide, 1.7 μm, 2.1 x 150 mm
Catalog No. NC9860929 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This BEH Glycan Separation Technology Column is optimized for UPLC systems and fluorescence detection (FLR) for separating released glycans of biopharmaceuticals as their 2-aminobenzamide (2-AB) derivatives. It offers superior component resolution for glycan structures, enabling scientists to characterize, identify, and quantitate oligosaccharides isolated from glycoproteins.
  • Optimized for hydrophilic interaction (HILIC) separation.
  • Features BEH hybrid particle technology.
  • Provides superior component resolution for glycans.
  • Stable at low pH and elevated temperatures.
  • Reduces analysis time and cost.
  • Compatible with fluorescence and MS detection.
  • Includes eCord Intelligent Chip Technology.
Catalog No. NC9860929 Supplier Waters Corp Supplier No. 186004742
This BEH Glycan Separation Technology Column is optimized for UPLC systems and fluorescence detection (FLR) for separating released glycans of biopharmaceuticals as their 2-aminobenzamide (2-AB) derivatives. It offers superior component resolution for glycan structures, enabling scientists to characterize, identify, and quantitate oligosaccharides isolated from glycoproteins.
  • Optimized for hydrophilic interaction (HILIC) separation.
  • Features BEH hybrid particle technology.
  • Provides superior component resolution for glycans.
  • Stable at low pH and elevated temperatures.
  • Reduces analysis time and cost.
  • Compatible with fluorescence and MS detection.
  • Includes eCord Intelligent Chip Technology.