Learn More
Waters Corp Waters XBridge VanGuard Cartridge, BEH Amide, 130Å, 2.5 μm
Catalog No. NC1535267 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Cartridges extend analytical column lifetime by removing particulate contamination. This cartridge protects 2.1 mm and 3.0 mm I.D. analytical columns with 2.5 μm sorbent particles, enhancing performance by removing impurities and maintaining UPLC, UHPLC, and HPLC separation efficiency.
  • Protects 2.1 mm and 3.0 mm I.D. analytical columns with 2.5 μm sorbent particles.
  • Simple setup; removes chemical and particle impurities.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Standalone device, links to UPLC Column intake.
  • Designed for UPLC columns in reversed-phase liquid chromatography.
  • Wide operational pH range of 2 to 11.
  • Withstands pressures up to 18,000 psi (1240 Bar).
  • QC tested for consistent performance and prolonged column life.
Catalog No. NC1535267 Supplier Waters Corp Supplier No. 186007763
VanGuard Cartridges extend analytical column lifetime by removing particulate contamination. This cartridge protects 2.1 mm and 3.0 mm I.D. analytical columns with 2.5 μm sorbent particles, enhancing performance by removing impurities and maintaining UPLC, UHPLC, and HPLC separation efficiency.
  • Protects 2.1 mm and 3.0 mm I.D. analytical columns with 2.5 μm sorbent particles.
  • Simple setup; removes chemical and particle impurities.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Standalone device, links to UPLC Column intake.
  • Designed for UPLC columns in reversed-phase liquid chromatography.
  • Wide operational pH range of 2 to 11.
  • Withstands pressures up to 18,000 psi (1240 Bar).
  • QC tested for consistent performance and prolonged column life.