Learn More
Waters Corp Column, Amide, 1.7 μm, 300 Å, 2.1 x 20 mm
Catalog No. NC3761177 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for robust and reliable separations of cell and gene therapeutics, including oligonucleotides, nucleic acids, and viral protein capsids. It utilizes hydrophilic interaction chromatography (HILIC) with an amide-bonded stationary phase.
  • MaxPeak High Performance Surface (HPS) reduces unwanted analyte interactions, improving peak shape, recovery, and reproducibility.
  • Chemically rugged, oligonucleotide batch-tested BEH particles ensure stringent quality control.
  • Supports HILIC and facilitates ion pairing-free mobile phase conditions through strong hydrogen bonding.
  • Manufactured in a cGMP, ISO 9001 certified plant under stringent protocols.
Catalog No. NC3761177 Supplier Waters Corp Supplier No. 186011248
This column is designed for robust and reliable separations of cell and gene therapeutics, including oligonucleotides, nucleic acids, and viral protein capsids. It utilizes hydrophilic interaction chromatography (HILIC) with an amide-bonded stationary phase.
  • MaxPeak High Performance Surface (HPS) reduces unwanted analyte interactions, improving peak shape, recovery, and reproducibility.
  • Chemically rugged, oligonucleotide batch-tested BEH particles ensure stringent quality control.
  • Supports HILIC and facilitates ion pairing-free mobile phase conditions through strong hydrogen bonding.
  • Manufactured in a cGMP, ISO 9001 certified plant under stringent protocols.