Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation ACQUITY UPLC BEH C18 Method Transfer Kit, 130Å, 1.7 to 5 μm, 2 pk

Catalog No. 50791262 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Method transfer kits are designed to maintain the integrity of a separation when transferring between UPLC and HPLC platforms. These kits provide a UPLC column with an HPLC column that has equivalent selectivity and resolving power, based on the concept of maintaining the column length to particle size ratio [L/dp]. A columns calculator can be used to fully transfer methods between HPLC and UPLC.

  • C18 chemistry.
  • Reversed phase separation mode.
  • Hybrid particle substrate.
  • pH range of 1 to 12.
  • Endcapped design.
  • Spherical particle shape.
  • Particle size from 1.7 to 5 μm.
  • Pore size of 130 Å.
  • Format as a method transfer kit.
  • Surface area of 185.
  • BEH particle technology.
  • USP classification L1.
  • Carbon load of 18%.
  • Includes 2 units per package.

Catalog No. 50-791-262 Supplier Waters Corp Supplier No. 186004958
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Method transfer kits are designed to maintain the integrity of a separation when transferring between UPLC and HPLC platforms. These kits provide a UPLC column with an HPLC column that has equivalent selectivity and resolving power, based on the concept of maintaining the column length to particle size ratio [L/dp]. A columns calculator can be used to fully transfer methods between HPLC and UPLC.

  • C18 chemistry.
  • Reversed phase separation mode.
  • Hybrid particle substrate.
  • pH range of 1 to 12.
  • Endcapped design.
  • Spherical particle shape.
  • Particle size from 1.7 to 5 μm.
  • Pore size of 130 Å.
  • Format as a method transfer kit.
  • Surface area of 185.
  • BEH particle technology.
  • USP classification L1.
  • Carbon load of 18%.
  • Includes 2 units per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.