Learn More
Waters Corp Waters XBridge Column, Protein BEH C4, 300Å, 2.5 μm
Catalog No. NC1661546 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH C4 Column, 300Å, 2.5 μm, 3 mm × 150 mm columns are optimized and QC tested with the MassPREP Protein Mix standard for protein separations. The 300Å pore size trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for various protein-based applications.
  • Optimized and QC tested with MassPREP Protein Mix standard for protein separations.
  • 300Å pore size trifunctionally bonded BEH particle.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. NC1661546 Supplier Waters Corp Supplier No. 186009134
The XBridge Protein BEH C4 Column, 300Å, 2.5 μm, 3 mm × 150 mm columns are optimized and QC tested with the MassPREP Protein Mix standard for protein separations. The 300Å pore size trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for various protein-based applications.
  • Optimized and QC tested with MassPREP Protein Mix standard for protein separations.
  • 300Å pore size trifunctionally bonded BEH particle.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.