Learn More
Waters Corp Waters Method Transfer Kit, HILIC, 130Å, 1.7 to 5 μm
Catalog No. 50791264 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Method transfer kits ensure separation integrity during transfer between UPLC and HPLC platforms. They provide UPLC and HPLC columns with equivalent selectivity and resolving power, based on maintaining the column length [L] to particle size [dp] ratio [L/dp]. A columns calculator facilitates full method transfer.
  • HILIC chemistry
  • Hydrophilic interaction (HILIC) separation mode
  • Hybrid particle substrate
  • pH range of 1 to 9
  • Not endcapped
  • Spherical particle shape
  • Particle size of 1.7 to 5 μm
  • Pore size of 130 Å
  • Kit format
  • Surface area of 185
  • BEH particle technology
  • USP classification L3
  • 2 units per package
  • Designed to preserve the integrity of a separation when transferred between UPLC and HPLC platforms
  • Allows methods to be fully transferred between HPLC and UPLC
Catalog No. 50-791-264 Supplier Waters Corp Supplier No. 186004960
Method transfer kits ensure separation integrity during transfer between UPLC and HPLC platforms. They provide UPLC and HPLC columns with equivalent selectivity and resolving power, based on maintaining the column length [L] to particle size [dp] ratio [L/dp]. A columns calculator facilitates full method transfer.
  • HILIC chemistry
  • Hydrophilic interaction (HILIC) separation mode
  • Hybrid particle substrate
  • pH range of 1 to 9
  • Not endcapped
  • Spherical particle shape
  • Particle size of 1.7 to 5 μm
  • Pore size of 130 Å
  • Kit format
  • Surface area of 185
  • BEH particle technology
  • USP classification L3
  • 2 units per package
  • Designed to preserve the integrity of a separation when transferred between UPLC and HPLC platforms
  • Allows methods to be fully transferred between HPLC and UPLC