Learn More
Waters Corp Waters Corporation Protein Standard Mix, BEH200 SEC, 5-Component, Lyophilized
Catalog No. 50466296 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BEH200 SEC Protein Standard Mix is a 5-component protein mixture for benchmarking ACQUITY UPLC BEH200 SEC, 1.7 μm Columns. It is delivered as a lyophilized powder for stability and can be directly solubilized with 500 μL of 100 mM Sodium Phosphate Buffer, pH 6.8.
  • Five-component protein mixture including Thyroglobulin, IgG, BSA, Myoglobin, and Uracil.
  • Designed for benchmarking UPLC BEH200 SEC, 1.7 μm columns, ensuring consistent performance.
  • Delivered as a lyophilized powder for enhanced stability and packaged in vials for direct solubilization.
  • Can be directly solubilized with 500 μL of 100 mM Sodium Phosphate Buffer, pH 6.8.
  • Manufactured under strict quality control criteria for batch-to-batch reproducibility across UPLC and HPLC applications.
Catalog No. 50-466-296 Supplier Waters Corp Supplier No. 186006518
The BEH200 SEC Protein Standard Mix is a 5-component protein mixture for benchmarking ACQUITY UPLC BEH200 SEC, 1.7 μm Columns. It is delivered as a lyophilized powder for stability and can be directly solubilized with 500 μL of 100 mM Sodium Phosphate Buffer, pH 6.8.
  • Five-component protein mixture including Thyroglobulin, IgG, BSA, Myoglobin, and Uracil.
  • Designed for benchmarking UPLC BEH200 SEC, 1.7 μm columns, ensuring consistent performance.
  • Delivered as a lyophilized powder for enhanced stability and packaged in vials for direct solubilization.
  • Can be directly solubilized with 500 μL of 100 mM Sodium Phosphate Buffer, pH 6.8.
  • Manufactured under strict quality control criteria for batch-to-batch reproducibility across UPLC and HPLC applications.