Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Behenic acid, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012513 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2513 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2513 Supplier MilliporeSigma Supelco Supplier No. 119095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Behenic acid is a saturated fatty acid with cholesterol-raising ability in humans.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 112-85-6
Physical Form Neat
Linear Formula CH3(CH2)20 COOH
MDL Number MFCD00002807
Melting Point 72°C to 80°C; 79°C to 81°C
Synonym Docosanoic acid
Type Analytical Standard
Quantity 5 g
Formula Weight 340.58
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.