Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Behenyl Alcohol, Spectrum™ Chemical
SDP

Catalog No. 18609038 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
10 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-038 10 g Amber Glass Bottle
18-609-039 100 g Amber Glass Bottle
18-609-037 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-609-038 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B285910GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 661-19-8
Molecular Weight (g/mol) 326.6 g/mol

Specifications

CAS Max % 1
Color White
Packaging Amber Glass Bottle
Quantity 10 g
Percent Purity 95%
Grade Reagent
Physical Form Powder or Waxy Solid
Chemical Name or Material Behenyl Alcohol

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.