Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Behenyl Alcohol (Technical), 80%, MP Biomedicals

Catalog No. ICN204154 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204154 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204154 Supplier MP Biomedicals, Inc Supplier No. 0520415480

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 661-19-8
Molecular Formula C22H46O
Molecular Weight (g/mol) 326.609
InChI Key NOPFSRXAKWQILS-UHFFFAOYSA-N
Synonym 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22
PubChem CID 12620
ChEBI CHEBI:31000
IUPAC Name docosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCO

Specifications

Quantity 100 g
Formula Weight 326.61
Grade Technical
Chemical Name or Material Behenyl Alcohol, 80%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.