Learn More
Medchemexpress LLC Behenyl linoleate | 204688-40-4 | >99.0% | 10 MG
SDP
Catalog No. 5000295165
Encompass_Preferred
Behenyl linoleate is a long-chain wax ester (Docosyl (9Z,12Z)-9,12-octadecadienoate) used primarily as an evaporation retardant and supplied for research use. The compound has molecular formula C40H76O2, molecular weight 589.03 g/mol, and is reported at high purity for laboratory applications.
  • Evaporation retardant for formulations and coatings.
  • Long-chain wax ester with hydrophobic properties.
  • High chemical purity (reported >99.0%).
  • Molecular formula C40H76O2 and molecular weight 589.03 g/mol.
  • Intended for research use; not for human or animal consumption.
Catalog No. 50-002-95165 Supplier Medchemexpress LLC Supplier No. HY17194710MG
Behenyl linoleate is a long-chain wax ester (Docosyl (9Z,12Z)-9,12-octadecadienoate) used primarily as an evaporation retardant and supplied for research use. The compound has molecular formula C40H76O2, molecular weight 589.03 g/mol, and is reported at high purity for laboratory applications.
  • Evaporation retardant for formulations and coatings.
  • Long-chain wax ester with hydrophobic properties.
  • High chemical purity (reported >99.0%).
  • Molecular formula C40H76O2 and molecular weight 589.03 g/mol.
  • Intended for research use; not for human or animal consumption.