Learn More
Siemens Healthineers Behring Coagulation Cups 500St
Catalog No. 23044699 Shop All Siemens Healthineers Products
Change view
Click to view available options
Capacity (Metric):
4 mL
Catalog No. Capacity (Metric)
23-044-699 4 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-699 Supplier Siemens Healthineers Supplier No. 10446526
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with the Dade™ Behring Coagulation Analysis System.
Behring Coagulation Cup serves as vessel for samples, human plasma and buffer solutions.
TRUSTED_SUSTAINABILITY
Specifications
Type Coagulation Sample Cups
For Use With (Equipment) Dade™ Behring Coagulation Analysis Systems
Capacity (Metric) 4 mL