Learn More
ATCC Streptococcus parasanguinis
Catalog No. 502380555
Encompass
Biovar 2 strain with the ability to produce aminopeptidase.
Catalog No. 50-238-0555 Supplier ATCC Supplier No. 903
Biovar 2 strain with the ability to produce aminopeptidase.