Learn More
Boca Scientific Inc Buccalyse Rapid Buccal Cell DNA Isolation Kit, 50 Extractions
Catalog No. NC1898702
Encompass
The Buccalyse Rapid Buccal Cell DNA Isolation Kit is a fast and simple one step, one tube method for extracting DNA from buccal swabs. The kit includes one solution that has been specifically formulated for use with buccal swabs. In only 20 minutes, this kit provides PCR-ready DNA that does not require centrifugation. The average total yield from an adult is 2-4ug.
Catalog No. NC1898702 Supplier Boca Scientific Inc Supplier No. BEK50
The Buccalyse Rapid Buccal Cell DNA Isolation Kit is a fast and simple one step, one tube method for extracting DNA from buccal swabs. The kit includes one solution that has been specifically formulated for use with buccal swabs. In only 20 minutes, this kit provides PCR-ready DNA that does not require centrifugation. The average total yield from an adult is 2-4ug.