Learn More
Medchemexpress LLC Bekanamycin (Kanamycin B) | 4696-76-8 | 250 MG
SDP
Catalog No. 5000414405
Encompass_Preferred
Bekanamycin (Kanamycin B) is an aminoglycoside antibiotic produced by *Streptomyces kanamyceticus*. It is effective against a range of Gram-positive and Gram-negative bacterial strains and serves as a precursor for semisynthetic antibiotics such as Arbekacin and Dibekacin.
  • Aminoglycoside antibiotic
  • Produced by *Streptomyces kanamyceticus*
  • Effective against Gram-positive bacterial strains
  • Effective against Gram-negative bacterial strains
  • Precursor for semisynthetic antibiotics
Catalog No. 50-004-14405 Supplier Medchemexpress LLC Supplier No. 250MGHYB1174
Bekanamycin (Kanamycin B) is an aminoglycoside antibiotic produced by *Streptomyces kanamyceticus*. It is effective against a range of Gram-positive and Gram-negative bacterial strains and serves as a precursor for semisynthetic antibiotics such as Arbekacin and Dibekacin.
  • Aminoglycoside antibiotic
  • Produced by *Streptomyces kanamyceticus*
  • Effective against Gram-positive bacterial strains
  • Effective against Gram-negative bacterial strains
  • Precursor for semisynthetic antibiotics