Learn More
MSC Bel-Art Polyethylene Tray, 12 x 16 x 3 in, without Faucet
Catalog No. 5000530706
Encompass
This general purpose tray is constructed from molded low-density polyethylene, offering a rugged, long-lasting, and corrosion-resistant solution for various laboratory and industrial applications. It features heavy walls and wide, top flanges for durability and can withstand temperatures up to 77°C (171°F). This specific model is supplied without a faucet and measures 12 x 16 x 3 inches.
  • Molded low-density polyethylene construction
  • Rugged, long-lasting, and corrosion-resistant
  • Heavy walls and wide, top flanges
  • Withstands temperatures up to 77°C (171°F)
Catalog No. 50-005-30706 Supplier MSC Supplier No. 74366808
This general purpose tray is constructed from molded low-density polyethylene, offering a rugged, long-lasting, and corrosion-resistant solution for various laboratory and industrial applications. It features heavy walls and wide, top flanges for durability and can withstand temperatures up to 77°C (171°F). This specific model is supplied without a faucet and measures 12 x 16 x 3 inches.
  • Molded low-density polyethylene construction
  • Rugged, long-lasting, and corrosion-resistant
  • Heavy walls and wide, top flanges
  • Withstands temperatures up to 77°C (171°F)