Learn More
MSC Bel-Art Rectangular Tank, Polyethylene, 10 gal, with Top Flanges, 18 x 13 x 10 in.
Catalog No. 5000515410
Encompass
The Bel-Art Heavy Duty Polyethylene Rectangular Tank is designed for laboratory and industrial use, offering a 10-gallon capacity. Made from rotational-molded polyethylene, this tank measures 18 x 13 x 10 inches and includes a lid, making it suitable for various storage and transfer applications.
  • Seamless, rotational-molded plastic tanks are lightweight and easy-to-handle.
  • Resistant to most acids and all alkalis.
  • Wide top flanges.
  • Includes covers.
  • Available with or without an optional needle type polyethylene faucet with 12.7 mm bore.
Catalog No. 50-005-15410 Supplier MSC Supplier No. 85798825
The Bel-Art Heavy Duty Polyethylene Rectangular Tank is designed for laboratory and industrial use, offering a 10-gallon capacity. Made from rotational-molded polyethylene, this tank measures 18 x 13 x 10 inches and includes a lid, making it suitable for various storage and transfer applications.
  • Seamless, rotational-molded plastic tanks are lightweight and easy-to-handle.
  • Resistant to most acids and all alkalis.
  • Wide top flanges.
  • Includes covers.
  • Available with or without an optional needle type polyethylene faucet with 12.7 mm bore.