Learn More
MSC Bel-Art Rectangular Tank, Polyethylene, 57 L, No Faucet, 1 ea
Catalog No. 5000793429
Encompass
This tank is a seamless, rotational-molded polyethylene plastic tank designed to be lightweight and easy to handle. It is resistant to most acids and all alkalis, featuring wide top flanges and including covers.
  • Seamless, rotational-molded polyethylene plastic construction
  • Lightweight and easy to handle
  • Resistant to most acids and all alkalis
  • Wide top flanges
  • Includes covers
Catalog No. 50-007-93429 Supplier MSC Supplier No. 85798833
This tank is a seamless, rotational-molded polyethylene plastic tank designed to be lightweight and easy to handle. It is resistant to most acids and all alkalis, featuring wide top flanges and including covers.
  • Seamless, rotational-molded polyethylene plastic construction
  • Lightweight and easy to handle
  • Resistant to most acids and all alkalis
  • Wide top flanges
  • Includes covers