Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil

Catalog No. 19821980 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (English):
13 to 20 gal.
2 to 4 gal.
20 to 30 gal.
40 to 55 gal.
5 to 9 gal.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-821-980 40 to 55 gal.
03-411-700 2 to 4 gal.
03-411-701 5 to 9 gal.
03-411-702 13 to 20 gal.
03-411-703 20 to 30 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-821-980 Supplier Bel-Art™ Supplier No. F131643848
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.

  • Bright red polypropylene bags include black biohazard symbol
  • Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
  • Sterilization patch darkens when subjected to steam sterilization
  • High temperature steam autoclavable to 135°C (275°F)

Specifications

Description 38W x 48H in. (97 x 122cm)
Autoclavable Autoclavable
Certifications/Compliance ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact
Closure Type Zip Tie
Color Red
Disposable Single-use
For Use With (Application) Used for biohazard waste
Includes Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish
Material Polypropylene
Sterility Non-sterile
Thickness (English) 1.50 mil
Dimensions (L x W) 38 x 48 in.
Length (English) 48 in.
Width (English) 38 in.
Height (English) 48 in.
Height (Metric) 122 cm
Product Type Biohazard Disposal Bag
Capacity (English) 40 to 55 gal.
Capacity (Metric) 175 L
Length (Metric) 122 cm
Width (Metric) 97 cm
Thickness (Metric) 0.279 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.