Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Bag Twist Tie Cord

Catalog No. 0181515 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Length (Metric):
457.2 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
01-815-15 457.2 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-815-15 Supplier Bel-Art™ Supplier No. H131900000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strong twist tie bag closures easier to use than string or rubber bands

  • Cut to optimum length and twist together for best closure performance
  • Use to bundle hard-to-handle items like rods, wires or tubes
  • Made of soft, copper-tinned, annealed iron wire sealed in extruded virgin vinyl
  • Autoclavable 121°C for 30 minutes
  • 1500 ft. per roll

Specifications

Type Ties
Material Nylon
Length (Metric) 457.2 m
Length (English) 1500 ft
For Use With (Application) Use to close bags or to package rods, wires and tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.