Learn More
Bel-Art™ Glove Box Accessory, Iris Port Entry
GSA_VA
Catalog No. 11391103 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Material:
Silicone Latex Rubber
Catalog No. Material
11-391-103 Silicone Latex Rubber
1 options
Catalog No. 11-391-103 Supplier Bel-Art™ Supplier No. H500250020
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Bare Hand Entry (Iris) Port allows bare hnd entry and operation.
  • Fits 7.75 in. I.D. (19.7 cm) glove ports
  • Made of ten layers of soft, conforming rubber
  • Easy access and withdrawal, plus excellent sealing
Order Info
One pair per pack.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Glove Box Port Cover
Material Silicone Latex Rubber
For Use With (Equipment) 19.7cm I.D. glove ports